missing translation for 'onlineSavingsMsg'
Learn More

PRIMA-1, MedChemExpress

Product Code. 16423927
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16423927

Brand: Medchem Express HY19980A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress PRIMA-1 (NSC-281668) is a mutant p53 reactivator, restores the sensitivity of TP53 mutant-type thyroid cancer cells to the histone methylation inhibitor 3-Deazaneplanocin A.

PRIMA-1 (NSC-281668) is a mutant p53 reactivator, restores the sensitivity of TP53 mutant-type thyroid cancer cells to the histone methylation inhibitor 3-Deazaneplanocin A.

Specifications

Chemical Name or Material PRIMA-1
CAS 5608-24-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C9H15NO3
Quantity 50 mg
Synonym NSC-281668
Solubility Information H2O : 100 mg/mL (539.90 mM; Need ultrasonic) ∣DMSO : 50 mg/mL (269.95 mM; Need ultrasonic)
SMILES O=C1C(CO)(CO)N2CCC1CC2
Molecular Weight (g/mol) 185.22
Formula Weight 185.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.