missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prilocaine, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Prilocaine
Specifications
Specifications
| Chemical Name or Material | Prilocaine |
| CAS | 721-50-6 |
| Analyte or Component Names | Prilocaine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H20 N2 O |
| Certifications/Compliance | ISO 17034 |
| Synonym | Prilocaine, (RS)-N-(2-Methylphenyl)-2-(propylamino)propanamide, Propanamide, N-(2-methylphenyl)-2-(propylamino)-, o-Propionotoluidide, 2-(propylamino)- (6CI,8CI), Astra 1512, Astra 1515, Citanest, N-(2,6-Dimethylphenyl)-2-(propylamino)propanamide, NSC 40027, Propitocaine, o-Methyl-2-propylaminopropionanilide, (+/-)-N-(2-Propylaminopropionyl)-2-toluidine, (+/-)-Prilocaine, 2-Methyl-alpha-propylaminopropionanilide, DL-(+/-)-Prilocaine |
| InChI Formula | InChI=1S/C13H20N2O/c1-4-9-14-11(3)13(16)15-12-8-6-5-7-10(12)2/h5-8,11,14H,4,9H2,1-3H3,(H,15,16) |
| SMILES | CCCNC(C)C(=O)Nc1ccccc1C |
| IUPAC Name | N-(2-methylphenyl)-2-(propylamino)propanamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction