missing translation for 'onlineSavingsMsg'
Learn More

Prilocaine Hydrochloride, TRC

Product Code. 30136319
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
Unit Size:
100mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136319 5 g 5g
30135542 1 g 1g
17964105 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136319 Supplier Toronto Research Chemicals Supplier No. TRCP7250005G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1786-81-8
Molecular Formula C13 H20 N2 O . Cl H
Molecular Weight (g/mol) 256.77
Synonym Prilocaine Hydrochloride, N-(2-Methylphenyl)-2-(propylamino)propanamide monohydrochloride, 2-(Propylamino)-o-propionotoluidide hydrochloride, Citanest Plain, Citanest hydrochloride, L 67, L 67 hydrochloride, Prilocaine chloride, Prilocaine hydrochloride, Propitocaine hydrochloride, Xylonest, α-(Propylamino)-2-methylpropionanilide hydrochloride
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
SMILES Cl.CCCNC(C)C(=O)Nc1ccccc1C

Specifications

InChI Formula InChI=1S/C13H20N2O.ClH/c1-4-9-14-11(3)13(16)15-12-8-6-5-7-10(12)2;/h5-8,11,14H,4,9H2,1-3H3,(H,15,16);1H
Quantity 5 g
Formula Weight 256.1342
Shipping Condition Room Temperature
Chemical Name or Material Prilocaine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.