missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Prilocaine Hydrochloride, Mikromol™
Description
Prilocaine Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Prilocaine Hydrochloride |
| CAS | 1786-81-8 |
| Analyte or Component Names | Prilocaine Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C13 H20 N2 O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Prilocaine Hydrochloride, N-(2-Methylphenyl)-2-(propylamino)propanamide monohydrochloride, 2-(Propylamino)-o-propionotoluidide hydrochloride, Citanest Plain, Citanest hydrochloride, L 67, L 67 hydrochloride, Prilocaine chloride, Prilocaine hydrochloride, Propitocaine hydrochloride, Xylonest, alpha-(Propylamino)-2-methylpropionanilide hydrochloride |
| InChI Formula | InChI=1S/C13H20N2O.ClH/c1-4-9-14-11(3)13(16)15-12-8-6-5-7-10(12)2;/h5-8,11,14H,4,9H2,1-3H3,(H,15,16);1H |
| SMILES | Cl.CCCNC(C)C(=O)Nc1ccccc1C |
| IUPAC Name | N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?