missing translation for 'onlineSavingsMsg'
Learn More

Prilocaine Hydrochloride, LoGiCal

Product Code. 30462032
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30462032 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30462032 Supplier LoGiCal Supplier No. LGCFOR0563.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Prilocaine Hydrochloride
CAS 1786-81-8
Analyte or Component Names Prilocaine Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C13 H20 N2 O . Cl H
Certifications/Compliance ISO 9001
Synonym Prilocaine Hydrochloride, N-(2-Methylphenyl)-2-(propylamino)propanamide monohydrochloride, 2-(Propylamino)-o-propionotoluidide hydrochloride, Citanest Plain, Citanest hydrochloride, L 67, L 67 hydrochloride, Prilocaine chloride, Prilocaine hydrochloride, Propitocaine hydrochloride, Xylonest, alpha-(Propylamino)-2-methylpropionanilide hydrochloride
InChI Formula InChI=1S/C13H20N2O.ClH/c1-4-9-14-11(3)13(16)15-12-8-6-5-7-10(12)2;/h5-8,11,14H,4,9H2,1-3H3,(H,15,16);1H
Solution Type Neat
SMILES Cl.CCCNC(C)C(=O)Nc1ccccc1C
IUPAC Name N-(2-methylphenyl)-2-(propylamino)propanamide;hydrochloride
Industry Type Clinical Testing
Molecular Weight (g/mol) 256.77
Quantity 10 mg
Formula Weight 256.1342 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.