missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pridinol Mesilate, Mikromol™
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
Description
Pridinol Mesilate
Specifications
Specifications
| Chemical Name or Material | Pridinol Methanesulfonate Salt |
| CAS | 6856-31-1 |
| Analyte or Component Names | Pridinol Methanesulfonate Salt |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N O . C H4 O3 S |
| Certifications/Compliance | ISO 17034 |
| Synonym | 3-(N-Piperidino)-1,1-diphenyl-1-propanol methanesulfonate, 3-Piperidino-1,1-diphenyl-1-propanol methanesulfonate, Konlax, Loxeen, Lyseen, Mitanoline, Pridinol mesylate, Pridinol methanesulfonate, alpha,alpha-Diphenyl-1-piperidinepropanol methanesulfonate (1:1), 1-Piperidinepropanol, alpha,alpha-diphenyl-, methanesulfonate (salt) (8CI,9CI), Methanesulfonic acid, compd. with alpha,alpha-diphenyl-1-piperidinepropanol (7CI) |
| InChI Formula | InChI=1S/C20H25NO.CH4O3S/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;1-5(2,3)4/h1-2,4-7,10-13,22H,3,8-9,14-17H2;1H3,(H,2,3,4) |
| SMILES | CS(=O)(=O)O.OC(CCN1CCCCC1)(c2ccccc2)c3ccccc3 |
| IUPAC Name | 1,1-diphenyl-3-piperidin-1-ylpropan-1-ol;methanesulfonic acid |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction