missing translation for 'onlineSavingsMsg'
Learn More

PRICKLE4 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Product Code. 18423658 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18423658 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18423658 Supplier Novus Biologicals Supplier No. NBP281939AF488

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRICKLE4 Polyclonal antibody specifically detects PRICKLE4 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE4
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 488
Formulation 50mM Sodium Borate
Gene Alias over-expressed breast tumor protein, prickle homolog 4 (Drosophila), prickle-like protein 4
Host Species Rabbit
Immunogen PRICKLE4 antibody was raised against an 18 amino acid synthetic peptide near the amino terminus of human PRICKLE4. The immunogen is located within the first 50 amino acids of PRICKLE4. Amino Acid Squence: WPHQEDSPKPQDPGPPAN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 29964
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.