missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRICKLE3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564879
This item is not returnable.
View return policy
Description
Immunogen sequence: LIFCSRACSLG SEPTAPGPSR RSWSAGPVTA PLAASTASFS AVKGASETTT KGTSTELAPA TGPEEPSRFL RGAPHRHSMP ELGLRSVPEP PPESPGQPNL RPDDSAFGRQ S Highest antigen sequence identity to the following orthologs - mouse 79%, rat 75%.
LIM domain only 6 is a three LIM domain-containing protein. The LIM domain is a cysteine-rich sequence motif that binds zinc atoms to form a specific protein-binding interface for protein-protein interactions.
Specifications
| PRICKLE3 | |
| Polyclonal | |
| Unconjugated | |
| Prickle3 | |
| LIM domain only 6; LIM domain only protein 6; LMO6; LMO-6; Pk3; prickle homolog 3; prickle homolog 3 (Drosophila); prickle planar cell polarity protein 3; Prickle3; Prickle-like protein 3; Sfc16; triple LIM domain protein 6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 4007 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O43900 | |
| Prickle3 | |
| Recombinant protein fragment of Human PRICKLE3 (Product #RP-91981) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction