missing translation for 'onlineSavingsMsg'
Learn More

PRICKLE2 Antibody, DyLight 650, Novus Biologicals™

Product Code. 18425046 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18425046 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18425046 Supplier Novus Biologicals Supplier No. NBP281938C

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRICKLE2 Polyclonal antibody specifically detects PRICKLE2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Formulation 50mM Sodium Borate
Gene Alias DKFZp686D143, EPM5, prickle homolog 2 (Drosophila), Prickle Planar Cell Polarity Protein 2, prickle-like protein 2
Host Species Rabbit
Immunogen PRICKLE2 antibody was raised against a 15 amino acid synthetic peptide near the center of human PRICKLE2. The immunogen is located within amino acids 420 - 470 of PRICKLE2. Amino Acid Squence: QPEMWGKHFSNPKRS
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 166336
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.