missing translation for 'onlineSavingsMsg'
Learn More

PRICKLE1 Antibody [DyLight 594], Novus Biologicals™

Product Code. 30761948 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30761948 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30761948 Supplier Novus Biologicals Supplier No. NBP346898DL594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PRICKLE1 Polyclonal antibody specifically detects PRICKLE1 in Human,Mouse samples. It is validated for ELISA,Immunoprecipitation,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen PRICKLE1
Applications ELISA, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias EPM1BMGC138902, FLJ31627, FLJ31937, prickle homolog 1 (Drosophila), prickle-like 1, prickle-like 1 (Drosophila), prickle-like protein 1, REST (RE-1 silencing transcription factor)/NRSF (neuron-restrictive silencerfactor)-interacting LIM domain protein, REST/NRSF-interacting LIM domain protein 1, RILPMGC138903
Host Species Rabbit
Immunogen prickle homolog 1(Drosophila). (Uniprot# Q96MT3)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 144165
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.