missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRH1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21460-1-AP-150UL
This item is not returnable.
View return policy
Description
This antibody specifically recognizes the 17 kDa protein.
The PRH1 gene encodes a member of the heterogeneous family of proline-rich salivary glycoproteins. The encoded preproprotein undergoes proteolytic processing to generate one or more mature isoforms before secretion from the parotid and submandibular/sublingual glands. In western population this locus is commonly biallelic and encodes proline-rich protein (PRP) isoforms, PRP-1 and PRP-2. The reference genome encodes the PRP-1 allele. Certain alleles of this gene are associated with susceptibility to dental caries. This gene is located in a cluster of closely related salivary proline-rich proteins on chromosome 12.Specifications
| PRH1 | |
| Polyclonal | |
| Unconjugated | |
| PRH1 | |
| Db F, Db s, Pa, Parotid acidic protein, Parotid double band protein, PIF F, PIF S, Pr1/Pr2, PRH1, PRH1/2, PRH2, Protein A Peptide P C, Protein C, PRP 1/PRP 2, PRP 3/PRP 4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 5554 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P02810 | |
| PRH1 | |
| PRH1 Fusion Protein Ag14833 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction