missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRG3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110217
This item is not returnable.
View return policy
Description
Immunogen sequence: LHLENDAPHL ESLETQADLG QDLDSSKEQE RDLALTEEVI QAEGEEVKAS ACQDNFEDEE AMESDPAALD KDFQCPREED IVEVQGSPRC KICRYLLVRT PKTFAEAQNV CSRCYGGNLV SIHDFNFNYR IQCCTSTVNQ AQVWIGGNLR GWFLWKRFCW TDGSHWNFAY WSPGQPGNGQ GSCVALCTKG GYWRRAQCDK QLPFVCSF.
Possesses similar cytotoxic and cytostimulatory activities to PRG2/MBP. In vitro, stimulates neutrophil superoxide production and IL8 release, and histamine and leukotriene C4 release from basophils. [UniProt].
Specifications
| PRG3 | |
| Polyclonal | |
| Unconjugated | |
| PRG3 | |
| eosinophil major basic protein 2; Eosinophil major basic protein homolog; major basic protein 2; MBP2; MBPH; Prepro-major basic protein homolog; prepro-MBPH; PRG3; proteoglycan 3; proteoglycan 3, pro eosinophil major basic protein 2; UNQ486/PRO1002 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 10394, 295706, 53856 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.69 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9JL95, Q9Y2Y8 | |
| PRG3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 18-225 of human PRG3 (NP_006084.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction