missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRG2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA530130
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, K562, THP-1, HL-60. Predicted reactivity: Human (99%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
The protein encoded by this gene is the predominant constituent of the crystalline core of the eosinophil granule. High levels of the proform of this protein are also present in placenta and pregnancy serum, where it exists as a complex with several other proteins including pregnancy-associated plasma protein A (PAPPA), angiotensinogen (AGT), and C3dg. This protein may be involved in antiparasitic defense mechanisms as a cytotoxin and helminthotoxin, and in immune hypersensitivity reactions. It is directly implicated in epithelial cell damage, exfoliation, and bronchospasm in allergic diseases.
Specifications
| PRG2 | |
| Polyclonal | |
| Unconjugated | |
| PRG2 | |
| BMPG; bone marrow proteoglycan; bone-marrow proteoglycan; EMBP; Eosinophil granule major basic protein; eosinophil major basic protein; LPR3; major basic protein 1; MBP; MBP1; Mbp-1; mMBP; mMBP-1; natural killer cell activator; Pregnancy-associated major basic protein; PRG2; PRG-2; proMBP; Proteoglycan 2; proteoglycan 2 preproprotein; proteoglycan 2, bone marrow; proteoglycan 2, bone marrow (natural killer cell activator, eosinophil granule major basic protein); proteoglycan 2, pro eosinophil major basic protein | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5553 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1.18 mg/mL | |
| PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
| P13727 | |
| PRG2 | |
| Recombinant protein encompassing a sequence within the center region of human PRG2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction