missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
PRG-1/LPPR4 Polyclonal antibody specifically detects PRG-1/LPPR4 in Human samples. It is validated for Western Blot, ELISA
Specifications
Specifications
| Antigen | PRG-1/LPPR4 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 5 - 10 mg/ml, ELISA |
| Formulation | PBS |
| Gene Alias | LPPR4 lipid phosphate phosphatase-related protein type 4, LPR4, PHP1, PRG1, PRG-1, RP4-788L13.1 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptide derived from the PRG-1/LPPR4 protein (Uniprot: Q7Z2D5) |
| Purification Method | Ammonium sulfate precipitation |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?