missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRG-1/LPPR4 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£175.00 - £401.00
Specifications
| Antigen | PRG-1/LPPR4 |
|---|---|
| Dilution | Western Blot 5 - 10 mg/ml, ELISA |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18314073
|
Novus Biologicals
NBP3-11909-25UG |
25 μg |
£175.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18347401
|
Novus Biologicals
NBP3-11909-100UG |
100 μg |
£401.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PRG-1/LPPR4 Polyclonal antibody specifically detects PRG-1/LPPR4 in Human samples. It is validated for Western Blot, ELISASpecifications
| PRG-1/LPPR4 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| LPPR4 lipid phosphate phosphatase-related protein type 4, LPR4, PHP1, PRG1, PRG-1, RP4-788L13.1 | |
| Synthetic peptide derived from the PRG-1/LPPR4 protein (Uniprot: Q7Z2D5) | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot 5 - 10 mg/ml, ELISA | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS | |
| 9890 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title