missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PREX2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117672
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Functions as a RAC1 guanine nucleotide exchange factor (GEF), activating Rac proteins by exchanging bound GDP for free GTP. Its activity is synergistically activated by phosphatidylinositol 3,4,5-trisphosphate and the beta gamma subunits of heterotrimeric G protein. Mediates the activation of RAC1 in a PI3K-dependent manner. May be an important mediator of Rac signaling, acting directly downstream of both G protein-coupled receptors and phosphoinositide 3-kinase.
Specifications
| PREX2 | |
| Polyclonal | |
| Unconjugated | |
| PREX2 | |
| 6230420N16Rik; AI316880; AI553603; C030045D06Rik; D430013K02; DEP domain containing 2; DEP domain-containing protein 2; DEP.2; Depdc2; phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 2 protein; phosphatidylinositol-3,4,5-trisphosphate dependent Rac exchange factor 2; phosphatidylinositol-3,4,5-trisphosphate-dependent Rac exchange factor 2; PPP1R129; Prex2; P-Rex2; protein phosphatase 1, regulatory subunit 129; ptdIns(3,4,5)-dependent Rac exchanger 2; si:ch211-278g15.1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 109294, 80243 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q3LAC4, Q70Z35 | |
| PREX2 | |
| Recombinant Human Phosphatidylinositol 3,4,5-trisphosphate-dependent Rac exchanger 2 protein (1417-1592aa). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction