Description
Use Thermo Scientific™ Pretreatment Cartridges and Filters when the quality of your feed water doesn't match the requirements for your Thermo Scientific™ Barnstead™ water purification system.
Use pretreatment when your feed water quality doesn't match the requirements for your system.
Specifications
Specifications
| For Use With (Equipment) | LabTower TII, Pacific RO, Pacific TII |
| Type | Pretreatment Cartridge |
| Includes | 5 μm Filter w/ Carbon Cartridge, Hardness Stabilizer Cartridge |