missing translation for 'onlineSavingsMsg'
Learn More

Pretomanid, MedChemExpress

Product Code. 16455156
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16455156 100 mg 100mg
16444198 10 mg 10mg
16420547 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16455156 Supplier Medchem Express Supplier No. HY10844

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Pretomanid (PA-824) is an antibiotic used for the research of multi-drug-resistant tuberculosis affecting the lungs. Pretomanid exhibits a sub-micromolar MIC against M. tuberculosis (MTB). The MIC values of PA-824 against a panel of MTB pan-sensitive and Rifampin mono-resistant clinical isolates range from 0.015 to 0.25 μg/mL.

Pretomanid (PA-824) is an antibiotic used for the research of multi-drug-resistant tuberculosis affecting the lungs. Pretomanid exhibits a sub-micromolar MIC against M. tuberculosis (MTB). The MIC values of PA-824 against a panel of MTB pan-sensitive and Rifampin mono-resistant clinical isolates range from 0.015 to 0.25 μg/mL.

Specifications

Chemical Name or Material Pretomanid
Physical Form Solid
CAS 187235-37-6
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C14H12F3N3O5
For Use With (Application) COVID-19-immunoregulation
Synonym PA-824 (S)-PA 824
Solubility Information DMSO : 125 mg/mL (347.94 mM; Need ultrasonic)
SMILES FC(F)(F)OC(C=C1)=CC=C1CO[C@@H]2COC3=NC([N+]([O-])=O)=CN3C2
Molecular Weight (g/mol) 359.26
Formula Weight 359.26
Percent Purity 98.36%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.