missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pretomanid, MedChemExpress
Description
Pretomanid (PA-824) is an antibiotic used for the research of multi-drug-resistant tuberculosis affecting the lungs. Pretomanid exhibits a sub-micromolar MIC against M. tuberculosis (MTB). The MIC values of PA-824 against a panel of MTB pan-sensitive and Rifampin mono-resistant clinical isolates range from 0.015 to 0.25 μg/mL.
Specifications
Specifications
| Chemical Name or Material | Pretomanid |
| Physical Form | Solid |
| CAS | 187235-37-6 |
| Color | Off-White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C14H12F3N3O5 |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | PA-824 (S)-PA 824 |
| Solubility Information | DMSO : 125 mg/mL (347.94 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?