Learn More
Julabo™ Pressure Sensor
Product Code. p-8013545
Click to view available options
Connection Type:
2-Fittings M24 x 1.5 Male
2-Fittings M30 x 1.5 Male
2-Fittings M38 x 1.5 Male
Unit Size:
Each
Product Code. 15264343
Brand: Julabo™ 8980771
Only null left
Add to basket
Add to basket
Specifications
Description Pressure Sensor
For Use With (Equipment) Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t Temperature Control System
Connection Type 2-Fittings M24 x 1.5 Male