Learn More
Julabo™ Pressure Sensor
Click to view available options
Connection Type:
2-Fittings M24 x 1.5 Male
2-Fittings M30 x 1.5 Male
2-Fittings M38 x 1.5 Male
Unit Size:
Each
Specifications
Specifications
| Description | Pressure Sensor |
| For Use With (Equipment) | Suitable for PRESTO A30, PRESTO A40, PRESTO A45, PRESTO A45t Temperature Control System |
| Connection Type | 2-Fittings M30 x 1.5 Male |