Learn More
Specac™ Pressure Gauge Kit to fit Cyclone™ & Tornado™ Gas Cells
Product Code. 15260318
Encompass_Preferred
Click to view available options
Type:
Pressure Gauge Kit
Product Code. 15260318
Brand: Specac™ GS24160
Pressure Gauge Kit for use with the Cyclone and Tornado Long Pathlength Gas Cells. Ideal for working at low and high pressures. Gauges can be specified for low pressures (vacuum to 15 psi) and high pressures (vacuum to 125 psi).
Specifications
Type Pressure Gauge Kit