missing translation for 'onlineSavingsMsg'
Learn More

Preso/FRMPD4 Antibody (809408) [mFluor Violet 610 SE], Novus Biologicals™

Product Code. 30733121 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30733121 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30733121 Supplier Novus Biologicals Supplier No. FAB7676MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Preso/FRMPD4 Monoclonal antibody specifically detects Preso/FRMPD4 in Human,Mouse samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Preso/FRMPD4
Applications Western Blot
Classification Monoclonal
Clone 809408
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias FERM and PDZ domain containing 4, FERM and PDZ domain-containing protein 4, MGC142260, MGC142262, PDZ domain containing 10, PDZD10, PDZK10
Host Species Mouse
Immunogen E. coli-derived recombinant human Preso/FRMPD4, Asn890-Ser1022, Accession # Q14CM0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9758
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.