missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Presenilin-2 Antibody (APS 26) [Janelia Fluor™ 669], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NB100-74513JF669
This item is not returnable.
View return policy
Description
Presenilin-2 Monoclonal antibody specifically detects Presenilin-2 in Human,Mouse,Primate,Rat samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Presenilin-2 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| AD3LP, AD4AD3L, AD5, E5-1, EC 3.4.23, EC 3.4.23.-, presenilin 2 (Alzheimer disease 4), presenilin-2, PS-2, PS2CMD1V, PSNL2, STM2STM-2 | |
| Synthetic peptide corresponding to residues C L(317) P Y D P E M E E D S Y D S F G E P(334) of human PS2. | |
| 0.2 mL | |
| Alzheimers Research, Apoptosis, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway | |
| 5664 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| APS 26 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Primate, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction