missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Presenilin-2 Antibody (APS 21), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-74512V
This item is not returnable.
View return policy
Description
Presenilin-2 Monoclonal specifically detects Presenilin-2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Presenilin-2 | |
| Monoclonal | |
| DyLight 405 | |
| AD3LP, AD4AD3L, AD5, E5-1, EC 3.4.23, EC 3.4.23.-, presenilin 2 (Alzheimer disease 4), presenilin-2, PS-2, PS2CMD1V, PSNL2, STM2STM-2 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Presenilin 2 (APS 21) | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| APS 21 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| PSEN2 | |
| Synthetic peptide corresponding to residues C(31) Q E G R Q G P E D G E N T A(45) of human PS2. | |
| 0.2 mL | |
| Alzheimers Research, Apoptosis, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway | |
| 5664 | |
| Human, Mouse, Rat, Primate | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur