missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Presenilin-2 Antibody (APS 21), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-74512AF594
This item is not returnable.
View return policy
Description
Presenilin-2 Monoclonal antibody specifically detects Presenilin-2 in Human, Mouse, Rat, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| Presenilin-2 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| AD3LP, AD4AD3L, AD5, E5-1, EC 3.4.23, EC 3.4.23.-, presenilin 2 (Alzheimer disease 4), presenilin-2, PS-2, PS2CMD1V, PSNL2, STM2STM-2 | |
| Synthetic peptide corresponding to residues C(31) Q E G R Q G P E D G E N T A(45) of human PS2. | |
| 0.2 mL | |
| Alzheimers Research, Apoptosis, Neurodegeneration, Neuroscience, Phospho Specific, Stem Cell Signaling Pathway | |
| 5664 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Western Blot | |
| APS 21 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction