missing translation for 'onlineSavingsMsg'
Learn More

Presenilin-1 Antibody, Alexa Fluor™ 647, Novus Biologicals™

Product Code. 18418624 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18418624 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18418624 Supplier Novus Biologicals Supplier No. NBP176792AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Presenilin-1 Polyclonal antibody specifically detects Presenilin-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen Presenilin-1
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias AD3EC 3.4.23.-, Alzheimer disease 3, EC 3.4.23, FAD, presenilin 1, Protein S182, PS-1, PS1presenilin-1, PSNL1, S182
Host Species Rabbit
Immunogen Antibody was raised against a 23 amino acid synthetic peptide from near the carboxy terminus of human presenilin1. The immunogen is located within amino acids 330 - 380 of Presenilin1. Amino Acid Squence: SEEWEAQRDSHLGPHRSTPESRA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cancer, Hematopoietic Stem Cell Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission, Stem Cell Markers, Stem Cell Signaling Pathway, Tumor Suppressors, Wnt Signaling Pathway
Primary or Secondary Primary
Gene ID (Entrez) 5663
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.