missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ preQ1-alkyne
GREENER_CHOICE

Product Code. 18796016
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
This item is not returnable. View return policy

Product Code. 18796016

Brand: Tocris Bioscience™ 7806/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

preQ1 with alkyne handle for click chemistry

preQ1-alkyne is a modified analog of the natural substrate prequeosine1 (preQ1). It contains alkyne handle that enables users to attach additional groups to preQ1 via Cu(I)-catalyzed azide-alkyne cycloaddition (CuAAC) click chemistry. It can be used to generate photoactivatable probes that can control CRISPR gene editing via the RNA-TAG method.

Other RNA-TAG toolbox compounds include preQ1-biotin ( Cat. No. 7804 )

Plasmids for expression of the E.coli TGT enzyme ( #138201 ) and for cloning an RNA of interest into a vector containing the recognition motif at the 3' end ( #138209 ) are available from Addgene.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-[6-[[(2-Amino-4,7-dihydro-4-oxo-3H-pyrrolo[2,3-d]pyrimidin-5-yl)methyl]amino]hexyl]-3-(2-propyn-1-yloxy)propanamide
CAS 2771096-76-3
Quantity 10 mg
Target CRISPR Reagents
Molecular Formula C19H28N6O3
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.