missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ PrepSEQ™ Rapid Spin Sample Prep Kit - Extra Clean & Bead Beating

Product Code. 12173911 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
100 preps
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12173911 100 preps Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12173911 Supplier Applied Biosystems™ Supplier No. 4468304

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

PrepSEQ™ Rapid Spin Sample Prep Kit - Extra Clean & Bead Beating

The PrepSEQ™ Rapid Spin Sample Prep Kit - Extra Clean & Bead Beating was developed for the AFNOR-approved MicroSEQ™ Listeria species and Listeria monocytogenes Real-Time PCR detection workflows. A spin, transfer, and bead beating protocol efficiently lyses Listeria from a 750μL aliquot of more challenging enriched food samples (e.g., samples containing high fat or otherwise prone to PCR inhibition). The PrepSEQ™ Rapid Spin Sample Prep Kit - Extra Clean & Bead Beating meets the needs of lower throughput food laboratories.

NOTE: Standard samples that do not contain high fat or demonstrate PCR inhibition should be processed using the PrepSEQ™ Rapid Spin Sample Prep Kit - Bead Beating.

  • Designed for Food Safety and Environmental Monitoring applications
  • Supports low-to-medium throughput (up to 100 samples a day)
  • Inactivates PCR inhibitors and produces high quality DNA for downstream detection
  • Uses a simple spin protocol

    Optimized for PCR-based Food Safety and Environmental Monitoring
    For laboratories involved in food safety and environmental monitoring, PCR-based methods are rapidly becoming the standard for accurate, sensitive bacterial detection. Use this sample preparation method for fast, cost-effective extraction of high-quality DNA from the most challenging sample types.

    Designed for Laboratories that Prepare up to 100 Samples a Day
    A spin column-based preparation enables small batch sizes to be managed while requiring only simple benchtop devices to process, thereby limiting the capital expenditure associated with much higher throughput automation devices.

    Removes PCR Inhibitors and Produces High Quality DNA
    The PrepSEQ™ Rapid Spin Sample Prep Kit - Extra Clean & Bead Beating offers a simple, cost-effective way to prepare Listeria DNA from more difficult starting matrices such as cheese, milk, and high fat meats. During the extraction process, the media inside the spin column removes PCR inhibitors from the sample, allowing the bacteria to pass through into the sample collection tube. The pellet is resuspended, then transferred (to avoid carryover, which inhibits PCR) to a new tube containing zirconium beads for mechanical lysis.
  • TRUSTED_SUSTAINABILITY

    Specifications

    Content And Storage Store lysis buffer in refrigerator (2–8°C). DO NOT FREEZE.
    High-throughput Compatibility High-throughput Compatible, Automated Protocols
    Kit Type Rapid Spin Sample Prep Kit
    Product Line PrepSEQ
    Quantity 100 preps
    For Use With (Application) Food and Beverage
    Purification Target Bacterial DNA
    Sample Type Food & Environmental Samples

    For Testing of Food and Environmental Samples Only.

    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.