missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PREPL Recombinant Rabbit Monoclonal Antibody (K01_2P02)

Product Code. 30830379
Change view
Click to view available options
Quantity:
50 μL
100 μL
Unit Size:
100µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30830379 50 μL -
30826317 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30830379 Supplier Invitrogen™ Supplier No. MA565631
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: KDGKLVPMTV FHKTDSEDLQ KKPLLVHVYG AYGMDLKMNF RPERRVLVDD GWILAYCHVR GGGELGLQWH ADGRLTKKLN GLADLEACIK TLHGQGFSQP SLTTLTAFSA GGVLAGALCN SNPELVRAVT LEAPFLDVLN TMMDTTLPLT LEELEEWGNP SSDEKHKNYI KRYCPYQNIK PQHYPSIHIT AYENDERVPL K Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 96%, Pig - 97%, Cynomolgus monkey - 97%.

The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PREPL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_2P02
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 0.05% BSA, 40% glycerol, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene Prepl
Gene Accession No. Q4J6C6, Q5HZA6, Q8C167
Gene Alias 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143
Gene Symbols Prepl
Host Species Rabbit
Immunogen Recombinant protein of human PPCEL/PREPL (aa 451-651).
Purification Method Protein A/G
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100762687, 213760, 298771, 9581
Target Species Human, Mouse, Rat, Hamster
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.