missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PREPL Recombinant Rabbit Monoclonal Antibody (JE65-64)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544739
This item is not returnable.
View return policy
Description
PREPL Recombinant Monoclonal Antibody for Western Blot, IHC (P)
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
Specifications
| PREPL | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q4J6C6, Q5HZA6, Q8C167 | |
| Prepl | |
| Recombinant protein within human PREPL aa 621-720/727. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE65-64 | |
| Unconjugated | |
| Prepl | |
| 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 213760, 298771, 9581 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction