missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PREPL Recombinant Rabbit Monoclonal Antibody (24GB995)
Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA557641
This item is not returnable.
View return policy
Description
PREPL Recombinant Monoclonal Antibody for Western Blot, Flow
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
Specifications
| PREPL | |
| Recombinant Monoclonal | |
| Unconjugated | |
| Prepl | |
| 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 213760, 298771, 9581 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 24GB995 | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q4J6C6, Q5HZA6, Q8C167 | |
| Prepl | |
| A synthesized peptide derived from human PREPL (aa 601-685). | |
| 20 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction