Learn More
Invitrogen™ PREPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589772
Description
Immunogen sequence: AVTLEAPFLD VLNTMMDTTL PLTLEELEEW GNPSSDEKHK NYIKRYCPYQ NIKPQHYPSI HITAYENDER VPLKGIVSYT EKLKEAIAEH AKDTGEGYQT PNIILDIQPG GNHVIEDSHK KITAQIKFLY EELGLDSTSV FEDLKKYLKF; Positive Samples: U-87MG, Mouse brain, Rat brain; Cellular Location: Cytoplasm, cytosol.
The protein encoded by this gene belongs to the prolyl oligopeptidase subfamily of serine peptidases. Mutations in this gene have been associated with hypotonia-cystinuria syndrome, also known as the 2p21 deletion syndrome.
Specifications
| PREPL | |
| Polyclonal | |
| Unconjugated | |
| Prepl | |
| 2810457N15Rik; 9530014L06Rik; D030028O16Rik; KIAA0436; mKIAA0436; Prepl; prolyl endopeptidase; prolyl endopeptidase-like; Prolylendopeptidase-like; putative prolyl oligopeptidase; RGD1310143 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 213760, 298771, 9581 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.42 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q4J6C6, Q5HZA6, Q8C167 | |
| Prepl | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 578-727 of human PREPL (NP_0060272). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.