missing translation for 'onlineSavingsMsg'
Learn More

BD Prepared Schaedler Agar with Vitamin K1 and 5 percent Sheep Blood Plated Media

Product Code. 10135395
Change view
Click to view available options
Unit Size:
Pack of 120
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10135395 Pack of 120
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10135395 Supplier BD Supplier No. 254084

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Certifications/Compliance CE, IVD
Content And Storage Store Plates in the Dark at 2 to 8°C
Detects Fastidious Strictly Anaerobic Bacteria
Format 90mm Plate
Media Type Non-selective Medium
Product Type Agar
Sterility Sterile
Formulation 8.2g/L Pancreatic Digest of Casein, 2.5g/L Peptic Digest of Animal Tissue, 1g/L Papaic Digest of Soybean Meal, 5.8g/L Glucose, 5g/L Yeast Extract, 1.7g/L Sodium Chloride, 0.8g/L Dipotassium Phosphate, 0.4g/L L-Cystine, 0.01g/L Hemin, 0.01g/L Vitamin K, 3g/L Tris (hydroxymethyl) aminomethane, 13.5g/L Agar, 5% Sheep Blood, defibrin ated, 7.6±0.2 pH, 1L Purified Water
For Use With (Application) For isolation and cultivation of Fastidious Strictly Anaerobic Bacteria from Clinical Specimens
Incubation Conditions Incubate for 48 to 72 hr. in an Anaerobic Atmosphere for 35 to 37°C
Incubation Time 48 to 72 hr.
Regulatory Status CE, IVD
Specimen Type Clinical Specimens
Diameter (Metric) 90 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.