missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Applied Biosystems™ Premix Buffer Concentrate, 100 mL
Shop All Applied Biosystems Products
Click to view available options
:
Store in refrigerator (2–8°C).
Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Specifications
| Chemical Name or Material | N-terminal Sequencing Reagent |
| Physical Form | Liquid |
| Quantity | 100 mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction