Learn More
Manutan Premium mobile truck, yellow, 118 litre capacity
Product Code. 12834659
Encompass_Preferred
Product Code. 12834659 Supplier Manutan Supplier No. A343986
Only null left
Add to basket
Add to basket
High quality trucks ideal for all industrial, hospitality and commercial based applications. Manufactured from hard wearing 100% food grade polyethylene, durable and easy to clean. Tapered sides to allow easy nesting with a wide operating temperature range of -20°C to +60°C. 320 litre and 445 litre trucks have reinforced plastic bases for additional support with heavy loads.
  • Yellow
  • Dimensions (H x W x D): 760mm x 460mm x 460mm
  • Load capacity: 100kg