Learn More
Vici Valco Instruments™ PEEK Loops for Rheodyne™ Valves
Product Code. 12750924
Change view
Click to view available options
Volume (Metric):
500 μL
Unit Size:
Each
Product Code. Volume (Metric) unitSize
12750924 500 μL Each
1 options
Product Code. 12750924 Supplier Vici Valco Instruments™ Supplier No. 724JRSLR2210
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
PEEK sample loops are particularly well suited for separations involving proteins, peptides, nucleic acids or other samples of biological origin as well as in IC systems where metal traces may interact with ionic samples or release transition metals that may deactivate ion exchange columns.
  • Fits all Rheodyne metal and metal-free valves
  • Wide range of volumes available
  • Biocompatible
  • Made of premium grade PEEK tubing
  • PEEK is a mechanically strong, chemically inert polymer ideal for HPLC applications.
  • Since our PEEK sample loops contain no metals or other extractable compounds, no foreign materials will be introduced into the system.
  • Softener-free polyethylene caps are used to avoid contamination of the tubing.
Specifications
Max. Pressure 3500 psi
Volume (Metric) 500 μL
Product Title
Välj ett problem
Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.