Learn More
Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector
Product Code. 12313850
Change view
Click to view available options
Tubing Size:
3/16 x 1/16 in.
5/32 x 1/16 in.
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tubing Size Unit Size
12313850 5/32 x 1/16 in. Pack of 10
12323850 3/16 x 1/16 in. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 12313850 Supplier Ark-Plas Products™ Supplier No. MAD80DF0
  • ONLINE EXCLUSIVE PRICE
    £17.00
    £0.66 / Pack of 10
    Save £16.34
    96% Off
In Stock
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Made from Kynar. Ark-Plas Products™ Ark-Plas Products™ Premium Grade Barbed Reducing Straight Connector is ideal for connecting tubes of various diameter. The connector offers easy fitting into tubing or hoses without the need for tools.
  • Made of Kynar
  • Economical straight barbed
  • Easily connect fluid paths without the use of tools
Specifications
Material Fluororesin
Tubing Size 5/32 x 1/16 in.
Color Natural
Product Type Straight reducing connector