missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ PRELP Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144877
This item is not returnable.
View return policy
Description
Immunogen sequence: GLRWINLDNN RIRKIDQRVL EKLPG Antigen sequence identity: Mouse 84%; Rat 96%.
PRELP is a leucine-rich repeat protein present in connective tissue extracellular matrix. This protein functions as a molecule anchoring basement membranes to the underlying connective tissue. This protein has been shown to bind type I collagen to basement membranes and type II collagen to cartilage. It also binds the basement membrane heparan sulfate proteoglycan perlecan. This protein is suggested to be involved in the pathogenesis of Hutchinson-Gilford progeria (HGP), which is reported to lack the binding of collagen in basement membranes and cartilage.
Specifications
| PRELP | |
| Polyclonal | |
| Unconjugated | |
| PRELP | |
| 55 kDa leucine-rich repeat protein of articular cartilage; 7330409J17Rik; MST161; MSTP161; PRELP; Prolargin; prolargin proteoglycan; proline and arginine rich end leucine rich repeat protein; proline arginine-rich end leucine-rich repeat; proline/arginine-rich end leucine-rich repeat protein; proline-arginine-rich end leucine-rich repeat protein; SLRR2A | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 5549 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P51888 | |
| PRELP | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 127-151. Recombinant protein control fragment (Product #RP-109763). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction