missing translation for 'onlineSavingsMsg'
Learn More

PRELP Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

£169.00 - £255.00

Specifications

Antigen PRELP
Dilution ELISA 1:5000-1:10000
Applications ELISA
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18614668
View Documents
Novus Biologicals
NBP3-00141-100ul
100 μg
£255.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18638837
View Documents
Novus Biologicals
NBP3-00141-50ul
50 μg
£169.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

PRELP Polyclonal antibody specifically detects PRELP in Human samples. It is validated for ELISA
Specifications

Specifications

PRELP
ELISA
Unconjugated
Rabbit
Human
55 kDa leucine-rich repeat protein of articular cartilage, MST161, MSTP161, prolargin, prolargin proteoglycan, proline arginine-rich end leucine-rich repeat protein, proline/arginine-rich end leucine-rich repeat protein, Proline-arginine-rich end leucine-rich repeat protein, SLRR2AMGC45323
Produced in rabbits immunized with purified, recombinant Human PRELP (Accession#: NP_002716.1; Gln21-Ile382)
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
ELISA 1:5000-1:10000
Polyclonal
Purified
RUO
0.2 um filtered solution in PBS
5549
IgG
Antigen and protein A Affinity-purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Datenschutzrichtlinie.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt