missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone, TRC

Product Code. 30136338
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30136338 5 g 5g
30137327 1 g 1g
17999174 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136338 Supplier Toronto Research Chemicals Supplier No. TRCP7122005G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 145-13-1
Molecular Formula C21 H32 O2
Molecular Weight (g/mol) 316.48
Synonym (3β)-3-Hydroxy-pregn-5-en-20-one, Pregn-5-en-20-one, 3β-hydroxy- (8CI), (3β)-3-Hydroxypregn-5-en-20-one, 17β-[1-Ketoethyl]-δ5-androsten-3β-ol, 3β-Hydroxy-5-en-20-one, 3β-Hydroxy-5-pregnene-20-one, 3β-Hydroxy-δ5-pregnen-20-one, 3β-Hydroxypregn-5-en-20-one, 5-Pregnenolone, Arthenolone, Bina-Skin, Enelone, NSC 1616, NSC 18158, Natolone, Pregn-5-en-3β-ol-20-one, Pregn-5-ene-3β-ol-20-one, Pregnelon, Pregnenolone, Pregnenolone (progesterone precursor), Pregnetan, Pregneton, Pregnolon, Prenolon, Regnosone, Skinostelon, δ5-Pregnen-3β-ol-20-one, δ5-Pregnene-3β-ol-20-one, δ5-Pregnenolone
IUPAC Name 1-[(3S,8S,9S,10R,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1
Formula Weight 316.2402
Percent Purity >95%
Chemical Name or Material Pregnenolone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.