missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone, MedChemExpress

Product Code. 16402069
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402069 1 g 1g
16490788 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16402069 Supplier Medchem Express Supplier No. HYB0151
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Pregnenolone (3β-Hydroxy-5-pregnen-20-one) is a powerful neurosteroid, the main precursor of various steroid hormones including steroid ketones.

Pregnenolone acts as a signaling-specific inhibitor of cannabinoid CB1 receptor, inhibits the effects of tetrahydrocannabinol (THC) that are mediated by the CB1 receptors. Pregnenolone can protect the brain from cannabis intoxication. Pregnenolone is also a TRPM3 channel activator, and also can weakly activate TRPM1 channels.

Specifications

Chemical Name or Material Pregnenolone
CAS 145-13-1
Color White
Purity Grade Notes Research
Molecular Formula C21H32O2
Quantity 1 g
Synonym 3β-Hydroxy-5-pregnen-20-one
Solubility Information DMSO : 25 mg/mL (78.99 mM; ultrasonic and warming and heat to 60°C) ∣H2O : 0.1 mg/mL (0.32 mM; Need ultrasonic)
SMILES C[C@@](C1=CC2)(CC[C@H](O)C1)[C@]3([H])[C@]2([H])[C@@](CC[C@@H]4C(C)=O)([H])[C@]4(C)CC3
Molecular Weight (g/mol) 316.48
Formula Weight 316.48
Percent Purity 98.0%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.