missing translation for 'onlineSavingsMsg'
Learn More

Pregnenolone (3beta-Hydroxypregn-5-en-20-one), Mikromol™

Product Code. 30051872
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30051872 25 mg 25mg
17997754 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051872 Supplier Mikromol Supplier No. MM0222.160025
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: A license is required to purchase this product; ROYAL DECREE OF 12 APRIL 1974.

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Pregnenolone (3beta-Hydroxypregn-5-en-20-one)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Pregnenolone
CAS 145-13-1
Analyte or Component Names Pregnenolone
Shipping Condition Room Temperature
Molecular Formula C21 H32 O2
Certifications/Compliance ISO 17025
Synonym 5-Pregnenolone, Arthenolone, Bina-Skin, Enelone, NSC 1616, NSC 18158, Natolone, Pregnelon, Pregnenolone, Pregnenolone (progesterone precursor), Pregnetan, Pregneton, Pregnolon, Prenolon, Regnosone, Skinostelon, (3beta)-3-Hydroxy-pregn-5-en-20-one, Pregn-5-en-20-one, 3beta-hydroxy- (8CI), (3beta)-3-Hydroxypregn-5-en-20-one, 17beta-[1-Ketoethyl]-Delta5-androsten-3beta-ol, 3beta-Hydroxy-5-en-20-one, 3beta-Hydroxy-5-pregnene-20-one, 3beta-Hydroxy-Delta5-pregnen-20-one, 3beta-Hydroxypregn-5-en-20-one, Pregn-5-en-3beta-ol-20-one, Pregn-5-ene-3beta-ol-20-one, Delta5-Pregnen-3beta-ol-20-one, Delta5-Pregnene-3beta-ol-20-one, Delta5-Pregnenolone
InChI Formula InChI=1S/C21H32O2/c1-13(22)17-6-7-18-16-5-4-14-12-15(23)8-10-20(14,2)19(16)9-11-21(17,18)3/h4,15-19,23H,5-12H2,1-3H3/t15-,16-,17+,18-,19-,20-,21+/m0/s1
SMILES CC(=O)[C@H]1CC[C@H]2[C@@H]3CC=C4C[C@@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
IUPAC Name 1-[(3S,8S,9S,10R,13S,14S,17S)-3-hydroxy-10,13-dimethyl-2,3,4,7,8,9,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl]ethanone
Molecular Weight (g/mol) 316.48
Quantity 25 mg
Formula Weight 316.2402 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.