missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Pregnanolone

Product Code. 18085292
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
This item is not returnable. View return policy

Product Code. 18085292

Brand: Tocris Bioscience™ 3652/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Postitive allosteric modulator of GABAA receptors

Pregnanolone is a GABAA receptor positive allosteric modulator. Neuroactive steroid.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3α,5β)-3-Hydroxy-pregnan-20-one
CAS 128-20-1
Quantity 50 mg
Target GABA-A Receptor Modulators
Molecular Formula C21H34O2
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.