missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pregnanediol-3-Glucuronide (PDG) Competitive ELISA Kit

Product Code. 17875307
Change view
Click to view available options
Quantity:
10 x 96 Tests
96 Tests
Unit Size:
Each
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17875307 10 x 96 Tests Pack of 10
15895853 96 Tests Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17875307 Supplier Invitrogen™ Supplier No. EIAPDGX10
  • ONLINE EXCLUSIVE PRICE
    £4085.00
    £3717.35 / Pack of 10
    Save £367.65
    9% Off
 Restricted Item
Estimated Shipment: 13-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

ELISA

The Pregnanediol-3-Glucuronide (PDG) ELISA quantitates PDG in extracted serμm/plasma, urine, dried fecal extracts or cell culture medium. The assay will exclusively recognize both natural and recombinant PDG. Principle of the method The PDG solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, binding to the target on a different epitope from the capture antibody. A conjugated enzyme has been incorporated into the assay. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is inversely proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Pregnanediol-3-Glucuronide (PDG) is a natural human metabolite of Pregnanediol generated in the liver by UDP glucuonyltransferase. Glucuronidation is used to assist in the excretion of toxic substances, drugs or other substances that cannot be used as an energy source. Glucuronic acid is attached via a glycosidic bond to the substance, and the resulting glucuronide, which has much higher water solubility than the original substance, is eventually excreted by the kidneys.
TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.391-50 ng/mL
Assay Sensitivity 0.18 ng/mL
Description Pregnanediol-3-Glucuronide (PDG) Competitive ELISA Kit
Assay Multiplex Assay on the Luminex Platform
Conjugate HRP
Detection Method Fluorescence
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Sample Type Fecal Extract, Plasma, Serum, Supernatant, Urine
Sufficient For 10 x 96 Assays
Target Species Chemical
For Use With (Application) ELISA, Protein Assays, Aktivity Assays, Protein Biology, Reproductive Biology, Inflammation, Oxidative Stress, Metabolism, Cell Signaling, Epigenetics, Hematology, DNA Damage, Cancer Biology, Immunology, Neuroscience, Neurobiology, Steroid Hormone Detecti
For Use With (Equipment) Absorbance Microplate Reader
Gene Alias C27H44O8; PDGC27H44O8; PDG;
Interassay CV 6.4%
Intraassay CV 4.1%
Kit Contents Pre-coated 96 well plate, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers, Standard, Assay Diluent concentrate, HRP-Conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
Label or Dye HRP
Protein Aliases PDG; C27H44O8
Quantity 10 x 96 Tests
Regulatory Status RUO
Research Discipline Reproductive Biology
Sample Volume Fecal Extract 50 μL, Plasma 50 μL, Serum 50 μL, Supernatant 50 μL, Urine 10 μL
Shipping Condition Wet or Dry Ice
Storage Requirements -20°C
Target Pregnanediol-3-Glucuronide
Test Time 1 hr 20 min
Total Assay Time 2 hr 30 min
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.