missing translation for 'onlineSavingsMsg'
Learn More

Pregnancy Zone Protein Antibody [Janelia Fluor™ 646], Novus Biologicals™

Product Code. 30734921 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30734921 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734921 Supplier Novus Biologicals Supplier No. AF8280J

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Goat Polyclonal Antibody

Pregnancy Zone Protein Polyclonal antibody specifically detects Pregnancy Zone Protein in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen Pregnancy Zone Protein
Applications Western Blot
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias C3 and PZP-like alpha-2-macroglobulin domain-containing protein 6, CPAMD6, pregnancy zone protein, pregnancy-zone protein
Host Species Goat
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human Pregnancy Zone Protein/PZP, Thr26-Val1482, Accession # P20742
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5858
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.