missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pregnancy specific beta 1 glycoprotein 11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£222.00 - £484.00
Specifications
| Antigen | Pregnancy specific beta 1 glycoprotein 11 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18451002
|
Novus Biologicals
NBP1-87795-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18264486
|
Novus Biologicals
NBP1-87795 |
0.1 mL |
£484.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pregnancy specific beta 1 glycoprotein 11 Polyclonal specifically detects Pregnancy specific beta 1 glycoprotein 11 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Pregnancy specific beta 1 glycoprotein 11 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| pregnancy specific beta-1-glycoprotein 11, pregnancy-specific beta-1-glycoprotein 11, Pregnancy-specific beta-1-glycoprotein 13, Pregnancy-specific glycoprotein 11, Pregnancy-specific glycoprotein 13, PS-beta-G-11, PS-beta-G-13, PSBG-11, PSBG-13, PSG13PSG14MGC22484 | |
| PSG11 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunofluorescence | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5680 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:STSLTIRVIAPPGLGTFAFNNPT | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title