missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uvex Preformed Mask silv-Air c 2110 FFP1
Shop All Uvex Products
Click to view available options
:
Filter Type: FFP1
Beskrivning
General Features
- Particle-filtering preformed mask with exhalation valve
- Color: white
Protection Features
- Certified FFP1 face mask to EN 149
- D label (dolomite dust test passed): maximum breathability even with really high dust levels
- Cannot be reused
Comfort Features
- Seamless headband and nose clip for a comfortable, secure fit
- Comfortable sealing lip in nose area for secure fit
- Exhalation valve to lessen exhalation resistance and to reduce heat and moisture inside the mask
Specifikationer
Specifikationer
| Certifications/Compliance (Europe) | EN 149 (D marked) |
| Color | White |
| Design Type | Cup/Molded |
| Disposable | Single-use |
| Filter Type | Particulate |
| Material | Polypropylene, Polyester |
| Product Type | General Purpose Particulate Respirator |
| Size | Standard |
| With Exhalation Valve | Yes |
| Filter Efficiency | FFP1 |
| Visa mer |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering