missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pref-1/DLK1/FA1, Rat anti-Mouse, Clone: PF105B, Novus Biologicals™
Rat Monoclonal Antibody
£280.00 - £425.00
Specifications
| Antigen | Pref-1/DLK1/FA1 |
|---|---|
| Clone | PF105B |
| Concentration | 1 mg/mL |
| Dilution | Western Blot, ELISA |
| Applications | Western Blot, ELISA |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18289927
|
Novus Biologicals
NBP2-80044-50UG |
50 μg |
£280.00
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18265817
|
Novus Biologicals
NBP2-80044-100UG |
100 μg |
£425.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Mouse samples. It is validated for Western Blot, ELISA.Specifications
| Pref-1/DLK1/FA1 | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rat | |
| Mouse | |
| delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG | |
| DLK1 | |
| IgG2a κ | |
| Protein G purified | |
| Recognizes mouse Pref-1/DLK1/FA1. Does not cross-react with human Pref-1/DLK1/FA1. |
| PF105B | |
| Western Blot, ELISA | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um-filtered solution in PBS, pH 7.4 | |
| 8788 | |
| Recombinant mouse Pref-1/DLK1/FA1 (extracellular domain). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title