missing translation for 'onlineSavingsMsg'
Learn More

Pref-1/DLK1/FA1, Mouse anti-Human, Clone: PF13-3, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-80046-50UG

 View more versions of this product

Product Code. 18295286

  • £301.00 / 50µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Pref-1/DLK1/FA1 Monoclonal specifically detects Pref-1/DLK1/FA1 in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, CyTOF-ready.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pref-1/DLK1/FA1
Monoclonal
1 mg/mL
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
delta-like 1 homolog (Drosophila), DLK, DLK-1, FA1fetal antigen 1, pG2delta-like homolog (Drosophila), preadipocyte factor 1, Pref-1, PREF1, protein delta homolog 1, secredeltin, ZOG
Mouse
Protein G purified
RUO
8788
Human
Purified
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry
PF13-3
Unconjugated
0.2 um-filtered solution in PBS, pH 7.4
DLK1
Recombinant human Pref-1/DLK1/FA1(extracellular domain).
50 μg
Primary
Recognizes human Pref-1/DLK1/FA1. Does not cross-react with mouse Pref-1/DLK1/FA1.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1 κ
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.